Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chst12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021528.3 | NP_067503.3 | 2 | 1358 | ||
GenBank mRNA | AJ289132.1 | - | 1 | 1271 | ||
AK142155.1 | - | 2 | 1356 | |||
AK163588.1 | - | 2 | 1329 | |||
AK170209.1 | - | 2 | 1321 | |||
AK170484.1 | - | 2 | 1325 | |||
AK171208.1 | - | 2 | 1325 | |||
BC003201.1 | - | 2 | 1339 | 3586766 |