Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ABLIM1 SIRNA Assays ›
Gene Symbol:
ABLIM1
Gene Name:
actin binding LIM protein 1
Gene Aliases:
ABLIM, LIMAB1, LIMATIN, abLIM-1
Chromosome Location:
Chr.10: 114431110 - 114821643 on Build GRCh38
UniGene ID:
Hs.438236
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s8187
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ABLIM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001003407.1
NP_001003407.1
9
1179
NM_001003408.1
NP_001003408.1
10
1263
NM_001322882.1
NP_001309811.1
10
1263
NM_001322883.1
NP_001309812.1
9
1179
NM_001322884.1
NP_001309813.1
9
978
NM_001322885.1
NP_001309814.1
9
978
NM_001322886.1
NP_001309815.1
9
978
NM_001322887.1
NP_001309816.1
9
1179
NM_001322888.1
NP_001309817.1
4
430
NM_001322889.1
NP_001309818.1
5
535
NM_001322890.1
NP_001309819.1
4
430
NM_001322891.1
NP_001309820.1
4
451
NM_001322892.1
NP_001309821.1
4
430
NM_001322893.1
NP_001309822.1
4
451
NM_001322894.1
NP_001309823.1
4
451
NM_001322895.1
NP_001309824.1
4
430
NM_001322898.1
NP_001309827.1
4
430
NM_001322899.1
NP_001309828.1
4
451
NM_002313.5
NP_002304.3
9
1156
NM_006720.3
NP_006711.3
5
514
XM_005269824.2
XP_005269881.1
9
1915
XM_005269826.1
XP_005269883.1
5
519
XM_005269827.4
XP_005269884.1
4
1149
XM_006717837.1
XP_006717900.1
10
1611
XM_006717846.3
XP_006717909.1
5
1254
XM_006717847.1
XP_006717910.1
4
348
XM_011539801.1
XP_011538103.1
10
1015
XM_011539802.1
XP_011538104.1
10
928
XM_011539803.1
XP_011538105.1
11
1033
XM_011539805.1
XP_011538107.1
10
1268
XM_017016242.1
XP_016871731.1
12
1117
XM_017016243.1
XP_016871732.1
12
1117
XM_017016244.1
XP_016871733.1
12
1117
XM_017016245.1
XP_016871734.1
10
1101
XM_017016246.1
XP_016871735.1
12
1117
XM_017016247.1
XP_016871736.1
9
897
XM_017016248.1
XP_016871737.1
8
813
XM_017016249.1
XP_016871738.1
12
1117
XM_017016250.1
XP_016871739.1
9
1017
XM_017016251.1
XP_016871740.1
11
1033
XM_017016252.1
XP_016871741.1
11
1033
XM_017016253.1
XP_016871742.1
11
1033
XM_017016254.1
XP_016871743.1
12
1117
XM_017016255.1
XP_016871744.1
9
1017
XM_017016256.1
XP_016871745.1
9
1915
XM_017016257.1
XP_016871746.1
11
1117
XM_017016258.1
XP_016871747.1
4
1149
XM_017016259.1
XP_016871748.1
4
348
GenBank mRNA
AF005654.1
-
9
1177
AK093471.1
-
10
783
AK098277.1
-
5
519
AK122891.1
-
10
1613
AK297344.1
-
9
1253
AK298709.1
-
4
334
AL710059.1
-
4
435
CR749819.1
-
4
426
D31883.2
-
9
981
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
cytoskeleton organization
visual perception
organ morphogenesis
lamellipodium assembly
cilium assembly
actin binding
protein binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0