Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jph2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205076.1 | NP_001192005.1 | 6 | 2393 | ||
NM_021566.2 | NP_067541.1 | 5 | 2473 | |||
GenBank mRNA | AB024447.1 | - | 5 | 2450 | ||
AK157145.1 | - | 6 | 2393 | |||
AY974567.1 | - | 2 | 215 |