Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk8ip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021921.3 | NP_068740.3 | 5 | 1984 | ||
GenBank mRNA | AF220195.1 | - | 5 | 1703 | ||
AF310135.1 | - | 5 | 1765 | |||
AK014339.1 | - | 5 | 1688 | |||
AK158968.1 | - | 5 | 1838 | |||
BC029704.1 | - | 5 | 1763 | 5359447 |