Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163028.1 | NP_001156500.1 | 6 | 925 | ||
NM_021486.3 | NP_067461.2 | 6 | 925 | |||
GenBank mRNA | AF271298.1 | - | 6 | 956 | ||
AF294899.1 | - | 6 | 938 | |||
AJ278064.2 | - | 6 | 937 | |||
AK054171.1 | - | 5 | 677 | |||
AK146472.1 | - | 6 | 923 | |||
BC125328.1 | - | 6 | 792 | 40129843 | ||
BC125330.1 | - | 6 | 792 | 40129845 | ||
BC145036.1 | - | 6 | 792 |