Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm20604 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142939.1 | NP_001136411.1 | 2 | 1351 | ||
GenBank mRNA | AK011927.1 | - | 2 | 1327 | ||
AK019599.1 | - | 2214 | ||||
BC014715.1 | - | 2 | 1300 | 4236212 | ||
BC055374.1 | - | 2 | 1304 | 30071431 |
Gene Symbol: Moap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142937.2 | NP_001136409.1 | 2 | 1200 | ||
NM_022323.7 | NP_071718.1 | 2 | 1244 | |||
GenBank mRNA | AF305551.1 | - | 1 | 975 |