Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptges | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022415.3 | NP_071860.1 | 3 | 317 | ||
GenBank mRNA | AB035323.1 | - | 3 | 269 | ||
AB041997.1 | - | 3 | 282 | |||
AK010757.1 | - | 3 | 282 | |||
AK033752.1 | - | 3 | 282 | |||
AK080897.1 | - | 3 | 322 | |||
AK150013.1 | - | 3 | 310 | |||
AK167015.1 | - | 3 | 186 | |||
AK172621.1 | - | 3 | 282 | |||
BC024960.1 | - | 3 | 276 | 4950950 |