Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRICKLE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001307979.1 | NP_001294908.1 | 9 | 1689 | ||
NM_006150.4 | NP_006141.2 | 9 | 1766 | |||
GenBank mRNA | AJ011654.1 | - | 9 | 1689 | ||
AK299407.1 | - | 7 | 1239 | |||
AK300866.1 | - | 8 | 1591 | |||
AK303200.1 | - | 9 | 1672 | |||
AK303308.1 | - | 8 | 1650 | |||
AK314788.1 | - | 9 | 1717 | |||
BC002468.2 | - | 9 | 1719 | 3347651 | ||
BC016856.1 | - | 9 | 1735 | 3846372 | ||
BT007423.1 | - | 9 | 1663 | |||
CR457211.1 | - | 9 | 1663 |