Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clstn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290989.1 | NP_001277918.1 | 7 | 1409 | ||
NM_023051.5 | NP_075538.1 | 8 | 1439 | |||
GenBank mRNA | AJ289016.1 | - | 8 | 1270 | ||
AK173059.1 | - | 7 | 1449 | |||
BC029027.1 | - | 3 | 524 | |||
BC053843.1 | - | 7 | 1378 | 6315355 | ||
CO433558.1 | - | 2 | 412 |