Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Twsg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023053.3 | NP_075540.1 | 5 | 694 | ||
GenBank mRNA | AF292033.1 | - | 633 | |||
AF295097.1 | - | 4 | 552 | |||
AJ297390.1 | - | 5 | 644 | |||
AK018524.1 | - | 1 | 61 | |||
AK033504.1 | - | 5 | 653 | |||
AK049646.1 | - | 5 | 688 | |||
AK203652.1 | - | 2 | 99 | |||
BC004850.1 | - | 5 | 649 | 2810960 |