Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eefsec | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023060.3 | NP_075547.1 | 2 | 461 | ||
GenBank mRNA | AF268871.1 | - | 2 | 425 | ||
AF283518.1 | - | 2 | 416 | |||
AK028424.1 | - | 2 | 443 | |||
AK087501.1 | - | 2 | 449 | |||
AK147924.1 | - | 2 | 430 | |||
AK171885.1 | - | 2 | 441 | |||
AK187190.1 | - | 1 | 63 | |||
AK207457.1 | - | 1 | 129 | |||
AK216395.1 | - | 1 | 63 | |||
BC024915.1 | - | 2 | 435 | 3158026 | ||
BC099675.1 | - | 2 | 419 | 5011896 |