Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ranbp17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023146.2 | NP_075635.1 | 23 | 2593 | ||
GenBank mRNA | AJ271458.1 | - | 23 | 2491 | ||
AK016583.1 | - | 23 | 2493 | |||
AK050150.1 | - | 19 | 2044 | |||
BC033503.1 | - | 24 | 2603 |