Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025316.2 | NP_079592.2 | 5 | 386 | ||
GenBank mRNA | AK002279.1 | - | 5 | 388 | ||
AK004372.1 | - | 5 | 386 | |||
AK005521.1 | - | 5 | 386 | |||
AK013223.1 | - | 5 | 388 | |||
AK018562.1 | - | 5 | 393 | |||
AK088061.1 | - | 5 | 395 | |||
BC025155.1 | - | 5 | 379 | 4024605 |