Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rmnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025343.5 | NP_079619.4 | 7 | 955 | ||
GenBank mRNA | AK007323.1 | - | 7 | 903 | ||
AK075603.1 | - | 7 | 938 | |||
AK161101.1 | - | 1 | 536 | |||
AK163523.1 | - | 191 | ||||
AK215071.1 | - | 1 | 19 | |||
BC019572.1 | - | 4 | 200 | |||
BC027299.1 | - | 7 | 903 | 3987394 | ||
BC036150.1 | - | 6 | 875 | 4977394 |