Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rmnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025343.5 | NP_079619.4 | 11 | 1388 | ||
GenBank mRNA | AK009625.1 | - | 2 | 176 | ||
AK075603.1 | - | 11 | 1371 | |||
AK163523.1 | - | 624 | ||||
AK205767.1 | - | 4 | 345 | |||
AK215625.1 | - | 4 | 324 | |||
BC019572.1 | - | 8 | 633 | |||
BC027299.1 | - | 11 | 1336 | 3987394 | ||
BC036150.1 | - | 10 | 1308 | 4977394 |