Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005577.2 | NP_005568.2 | 39 | 6008 | ||
GenBank mRNA | X06290.1 | - | 13457 |