Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025379.2 | NP_079655.1 | 1, 2 | 178 | ||
GenBank mRNA | AK002593.1 | - | 1, 2 | 125 | ||
AK003414.1 | - | 1, 2 | 129 | |||
AK132086.1 | - | 1, 2 | 174 | |||
BC024350.1 | - | 1, 2 | 119 | 4987472 | ||
BC089538.1 | - | 1, 2 | 178 | 6533206 |