Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxx1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024170.2 | NP_077132.1 | 1 | 884 | ||
GenBank mRNA | AK002323.1 | - | 1 | 820 | ||
BC003436.1 | - | 1 | 803 | 2647372 |
Gene Symbol: Cxx1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018063.1 | NP_001018073.1 | 1 | 852 | ||
GenBank mRNA | AK003646.1 | - | 1 | 831 | ||
AK206042.1 | - | 1 | 220 | |||
BC029838.1 | - | 1 | 852 | 3967024 |