Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025399.3 | NP_079675.1 | 2, 3 | 183 | ||
GenBank mRNA | AK003991.1 | - | 2, 3 | 190 | ||
AK013174.1 | - | 2, 3 | 139 | |||
AK184511.1 | - | 1, 2 | 61 | |||
BC025444.1 | - | 2, 3 | 185 | 3986887 |