Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025401.3 | NP_079677.1 | 3, 4 | 1738 | ||
GenBank mRNA | AK002730.1 | - | 3, 4 | 247 | ||
AK003992.1 | - | 3, 4 | 254 | |||
AK007726.1 | - | 3, 4 | 219 | |||
AK009854.1 | - | 3, 4 | 254 | |||
AK012803.1 | - | 3, 4 | 236 | |||
AK051149.1 | - | 3, 4 | 1739 | |||
AK154036.1 | - | 2, 3 | 2330 | |||
BC028498.1 | - | 3, 4 | 208 | 1400865 |