Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025401.3 | NP_079677.1 | 3 | 1664 | ||
GenBank mRNA | AK002730.1 | - | 3 | 173 | ||
AK003992.1 | - | 3 | 180 | |||
AK007726.1 | - | 3 | 145 | |||
AK009854.1 | - | 3 | 180 | |||
AK012803.1 | - | 3 | 162 | |||
AK051149.1 | - | 3 | 1665 | |||
AK154036.1 | - | 2 | 2256 | |||
BC028498.1 | - | 3 | 134 | 1400865 |