Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pithd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025411.4 | NP_079687.3 | 1, 2 | 311 | ||
GenBank mRNA | AF400670.1 | - | 1, 2 | 274 | ||
AK004208.1 | - | 1, 2 | 312 | |||
AK050261.1 | - | 1, 2 | 284 | |||
BC052695.1 | - | 1, 2 | 211 | 5715038 | ||
BY044314.1 | - | 1, 2 | 312 |