Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lce1m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025420.2 | NP_079696.1 | 2 | 691 | ||
GenBank mRNA | AK003825.1 | - | 2 | 687 | ||
AK004318.1 | - | 2 | 693 | |||
AK019517.1 | - | 2 | 691 | |||
BC109181.1 | - | 2 | 689 | 40049866 | ||
BC109182.1 | - | 2 | 693 | 40049867 |