Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Llph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025431.2 | NP_079707.1 | 3 | 424 | ||
GenBank mRNA | AK004503.1 | - | 3 | 424 | ||
AK007375.1 | - | 3 | 407 | |||
AK158700.1 | - | 3 | 402 | |||
BC024098.1 | - | 3 | 425 | 5148362 | ||
BC096681.1 | - | 3 | 423 | 1332438 | ||
BC106103.1 | - | 407 | 3661552 |