Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl7l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025433.3 | NP_079709.2 | 3 | 189 | ||
GenBank mRNA | AK005264.1 | - | 3 | 180 | ||
AK007884.1 | - | 3 | 178 | |||
AK008672.1 | - | 3 | 180 | |||
AK090038.1 | - | 3 | 189 | |||
AK090082.1 | - | 3 | 189 | |||
AK152288.1 | - | 2 | 118 | |||
AK166499.1 | - | 3 | 193 | |||
AK167036.1 | - | 3 | 188 | |||
AK212340.1 | - | 2 | 162 | |||
BC030165.1 | - | 3 | 187 | 5027519 |