Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thoc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285780.1 | NP_001272709.1 | 5 | 521 | ||
NM_025435.3 | NP_079711.1 | 4 | 443 | |||
GenBank mRNA | AK005176.1 | - | 4 | 370 | ||
AK007440.1 | - | 4 | 349 | |||
AK010958.1 | - | 4 | 384 | |||
AK013319.1 | - | 4 | 366 | |||
AK143106.1 | - | 2 | 1591 | |||
AK147003.1 | - | 5 | 424 | |||
BC027113.1 | - | 3 | 1391 | |||
BC042209.1 | - | 4 | 370 | |||
BC054419.1 | - | 4 | 403 | 4010343 | ||
BC116919.1 | - | 4 | 408 | 40126238 | ||
BC116945.1 | - | 4 | 408 | 40126264 | ||
CK329337.1 | - | 4 | 444 |