Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dimt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025447.4 | NP_079723.1 | 11 | 913 | ||
GenBank mRNA | AK007759.1 | - | 11 | 865 | ||
AK011544.1 | - | 11 | 870 | |||
AK015145.1 | - | 11 | 862 | |||
AK078186.1 | - | 9 | 1707 | |||
AK078401.1 | - | 9 | 1729 | |||
AK146188.1 | - | 11 | 884 | |||
AK161333.1 | - | 11 | 865 | |||
BC019799.1 | - | 11 | 886 | 5053500 |