Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290706.1 | NP_001277635.1 | 4 | 617 | ||
NM_025452.4 | NP_079728.2 | 4 | 635 | |||
NM_133881.1 | NP_598642.1 | 3 | 575 | |||
GenBank mRNA | AK007525.1 | - | 3 | 491 | ||
AK008935.1 | - | 4 | 550 | |||
AK009830.1 | - | 3 | 471 | |||
BC019563.1 | - | 4 | 564 | 4219997 |