Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6v1g1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024173.2 | NP_077135.1 | 3 | 400 | ||
GenBank mRNA | AB076406.1 | - | 3 | 297 | ||
AK003563.1 | - | 3 | 324 | |||
AK007596.1 | - | 3 | 343 | |||
AK010131.1 | - | 3 | 338 | |||
AK081611.1 | - | 3 | 360 | |||
AK088712.1 | - | 3 | 332 | |||
AK152699.1 | - | 3 | 324 | |||
AK159558.1 | - | 3 | 324 | |||
AK168972.1 | - | 3 | 338 | |||
AK169079.1 | - | 3 | 621 | |||
AK205668.1 | - | 1 | 16 | |||
BC003429.1 | - | 3 | 339 | 2646354 | ||
BC088836.1 | - | 3 | 293 | 6484999 | ||
CN838691.1 | - | 3 | 403 |