Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSAMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318915.1 | NP_001305844.1 | 2 | 881 | ||
NM_002338.4 | NP_002329.2 | 2 | 881 | |||
XM_011512840.2 | XP_011511142.1 | 2 | 885 | |||
XM_017006383.1 | XP_016861872.1 | 2 | 885 | |||
GenBank mRNA | AK299851.1 | - | 696 | |||
AK315286.1 | - | 2 | 820 | |||
BC033803.1 | - | 2 | 874 | 5188760 | ||
U41901.1 | - | 2 | 374 |