Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6v1c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025494.3 | NP_079770.2 | 11, 12 | 1131 | ||
GenBank mRNA | AB088407.1 | - | 11, 12 | 1078 | ||
AK006323.1 | - | 11, 12 | 1082 | |||
AK075779.1 | - | 11, 12 | 1103 | |||
AK129007.1 | - | 9 | 2653 | |||
AK151237.1 | - | 11, 12 | 1092 | |||
AK168914.1 | - | 11, 12 | 1082 | |||
BC010217.1 | - | 11, 12 | 1079 | 3498388 | ||
U13839.1 | - | 11, 12 | 1074 |