Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025517.3 | NP_079793.2 | 10 | 1156 | ||
GenBank mRNA | AK002543.1 | - | 10 | 1066 | ||
AK009245.1 | - | 10 | 1092 | |||
AK151108.1 | - | 10 | 1044 | |||
AK151400.1 | - | 10 | 1044 | |||
AK152129.1 | - | 10 | 1157 | |||
AK159961.1 | - | 10 | 1046 | |||
AK167830.1 | - | 10 | 1033 | |||
AK190960.1 | - | 4 | 312 | |||
AK193515.1 | - | 4 | 293 | |||
AK209245.1 | - | 4 | 293 | |||
AK212568.1 | - | 4 | 246 | |||
BC042473.1 | - | 1 | 347 | |||
BC051062.1 | - | 1 | 347 | |||
BC090403.1 | - | 10 | 1127 | 5293337 |