Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iscu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025526.4 | NP_079802.1 | 5 | 758 | ||
GenBank mRNA | AK009021.1 | - | 5 | 758 | ||
AK009421.1 | - | 5 | 762 | |||
AK039834.1 | - | 5 | 800 | |||
AK083247.1 | - | 5 | 766 | |||
BC028800.1 | - | 5 | 736 | |||
BC048409.1 | - | 5 | 755 | 6484953 | ||
BC096049.1 | - | 5 | 756 | 3466271 |