Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025539.2 | NP_079815.2 | 2 | 102 | ||
GenBank mRNA | AK009933.1 | - | 2 | 100 | ||
AK132532.1 | - | 103 | ||||
BC025153.1 | - | 1 | 78 | 4236805 |