Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aptx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025444.3 | NP_001020615.1 | 2, 3 | 199 | ||
NM_025545.4 | NP_079821.3 | 2, 3 | 334 | |||
GenBank mRNA | AK005286.1 | - | 1, 2 | 151 | ||
AK077351.1 | - | 2, 3 | 335 | |||
AK088928.1 | - | 2, 3 | 169 | |||
AY040780.1 | - | 2, 3 | 345 | |||
AY208844.1 | - | 2, 3 | 346 | |||
BB860888.1 | - | 2, 3 | 203 | |||
BC021872.1 | - | 2, 3 | 192 | 3994375 | ||
BC071213.1 | - | 2, 3 | 186 |