Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufa7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023202.4 | NP_075691.1 | 3 | 256 | ||
GenBank mRNA | AK002476.1 | - | 3 | 237 | ||
AK003468.1 | - | 3 | 237 | |||
AK010299.1 | - | 3 | 214 | |||
BC052817.1 | - | 3 | 220 | 6487401 | ||
BC055698.1 | - | 3 | 221 | 6411262 | ||
BX518947.1 | - | 2 | 133 | |||
KJ650204.1 | - | 3 | 198 |