Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usmg5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023211.4 | NP_075700.2 | 3 | 183 | ||
GenBank mRNA | AJ290947.1 | - | 191 | |||
AK008345.1 | - | 3 | 184 | |||
AK008511.1 | - | 1013 | ||||
AK038457.1 | - | 3 | 190 | |||
AK088856.1 | - | 3 | 201 | |||
BC024355.1 | - | 3 | 264 | |||
BC087919.1 | - | 3 | 183 | 6545418 |