Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exoc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025588.2 | NP_079864.1 | 8 | 963 | ||
GenBank mRNA | AK016532.1 | - | 9 | 1090 | ||
AK050068.1 | - | 8 | 971 | |||
BC048154.1 | - | 8 | 963 | 6400932 | ||
BC049102.1 | - | 8 | 963 | 5702581 |