Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asrgl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025610.3 | NP_079886.2 | 7 | 827 | ||
GenBank mRNA | AK006147.1 | - | 7 | 863 | ||
AK010381.1 | - | 7 | 869 | |||
AK030209.1 | - | 7 | 828 | |||
BC016106.1 | - | 5 | 579 | 4921265 |