Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubxn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024432.2 | NP_077752.1 | 9 | 1069 | ||
GenBank mRNA | AF272895.1 | - | 9 | 1084 | ||
AK009117.1 | - | 8 | 851 | |||
AK075705.1 | - | 7 | 2133 | |||
AK082510.1 | - | 1108 | ||||
AK131187.1 | - | 3 | 2841 | |||
AK148170.1 | - | 1037 | ||||
AK161273.1 | - | 9 | 1090 | |||
AK188370.1 | - | 1 | 32 | |||
BC016412.1 | - | 9 | 998 | 4215883 | ||
BC049963.1 | - | 9 | 1069 | 2655935 |