Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026844.3 | NP_081120.2 | 3 | 244 | ||
GenBank mRNA | AK002429.1 | - | 3 | 244 | ||
AK004189.1 | - | 4 | 436 | |||
AK009995.1 | - | 3 | 275 | |||
AK010073.1 | - | 3 | 275 | |||
AK013475.1 | - | 3 | 241 | |||
BC027549.1 | - | 3 | 214 | 1548292 |