Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpifb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025631.3 | NP_079907.2 | 8 | 754 | ||
GenBank mRNA | AK009346.1 | - | 8 | 727 | ||
AK009504.1 | - | 8 | 729 | |||
AK009619.1 | - | 8 | 727 | |||
AK009754.1 | - | 4 | 330 | |||
AK009776.1 | - | 8 | 711 | |||
AK009794.1 | - | 8 | 725 | |||
AK009871.1 | - | 8 | 729 | |||
AK010130.1 | - | 8 | 711 | |||
AK010154.1 | - | 8 | 725 | |||
AK010215.1 | - | 8 | 725 | |||
AK028156.1 | - | 8 | 725 | |||
AK028158.1 | - | 8 | 728 | |||
AK075874.1 | - | 8 | 725 | |||
AK160516.1 | - | 8 | 725 | |||
BC116681.1 | - | 8 | 709 | 40106489 | ||
BC116727.1 | - | 8 | 709 | 40106498 |