Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crls1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024385.1 | NP_001019556.1 | 3, 4 | 738 | ||
NM_025646.3 | NP_079922.2 | 3, 4 | 425 | |||
GenBank mRNA | AK002408.1 | - | 3, 4 | 740 | ||
AK004215.1 | - | 3, 4 | 454 | |||
AK016165.1 | - | 3, 4 | 426 | |||
AK017718.1 | - | 2, 3 | 587 | |||
BC048702.1 | - | 3, 4 | 719 | 6506241 | ||
BC086619.1 | - | 3, 4 | 695 |