Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Farsa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025648.3 | NP_079924.2 | 4, 5 | 557 | ||
GenBank mRNA | AK002577.1 | - | 4, 5 | 534 | ||
AK031697.1 | - | 4, 5 | 535 | |||
AK044637.1 | - | 4, 5 | 535 | |||
AK084031.1 | - | 4, 5 | 519 | |||
AK154629.1 | - | 4, 5 | 555 | |||
AK171872.1 | - | 4, 5 | 438 | |||
AK213018.1 | - | 21 | ||||
BC006862.1 | - | 4, 5 | 528 | 3595108 | ||
BC013533.1 | - | 4, 5 | 526 | 4238911 | ||
CT010350.1 | - | 4, 5 | 508 |