Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sike1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025679.3 | NP_079955.1 | 3 | 375 | ||
GenBank mRNA | AK012679.1 | - | 3 | 361 | ||
AK014113.1 | - | 3 | 376 | |||
AK014247.1 | - | 3 | 382 | |||
AK017452.1 | - | 3 | 375 | |||
AK017694.1 | - | 3 | 371 | |||
AK019264.1 | - | 3 | 361 | |||
AK033553.1 | - | 3 | 379 | |||
AK035590.1 | - | 3 | 389 | |||
AK038295.1 | - | 1 | 1562 | |||
BC004090.1 | - | 3 | 380 | 3593790 |