Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spryd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025697.3 | NP_079973.1 | 3 | 501 | ||
GenBank mRNA | AK018044.1 | - | 3 | 379 | ||
AK018153.1 | - | 3 | 502 | |||
AK018290.1 | - | 3 | 351 | |||
AK137685.1 | - | 350 | ||||
AK150789.1 | - | 3 | 376 | |||
AK167306.1 | - | 3 | 343 | |||
AK169059.1 | - | 3 | 350 | |||
BC037015.1 | - | 3 | 335 | 5344409 |