Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025706.3 | NP_079982.3 | 14 | 1599 | ||
GenBank mRNA | AK014477.1 | - | 14 | 1531 | ||
AK014795.1 | - | 9 | 982 | |||
AK078875.1 | - | 14 | 1545 | |||
AK135905.1 | - | 10 | 1075 | |||
AK147085.1 | - | 14 | 1562 | |||
AK162115.1 | - | 3 | 197 | |||
BC032087.1 | - | 14 | 1527 | |||
BC053395.1 | - | 14 | 1580 | 30054233 | ||
DQ054831.1 | - | 14 | 1530 |