Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aspn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172481.1 | NP_001165952.1 | 2 | 376 | ||
NM_025711.3 | NP_079987.2 | 2 | 393 | |||
GenBank mRNA | AF316825.1 | - | 1 | 113 | ||
AK014504.1 | - | 2 | 349 | |||
AK021386.1 | - | 2 | 348 | |||
AK085920.1 | - | 2 | 348 | |||
BC034888.1 | - | 2 | 295 | 1365428 | ||
BC145904.1 | - | 2 | 291 | 40131207 | ||
BC145930.1 | - | 2 | 274 | 40131233 |