Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rexo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025852.3 | NP_080128.2 | 4 | 2314 | ||
GenBank mRNA | AK173108.1 | - | 4 | 2279 | ||
BC049901.1 | - | 1 | 117 | |||
BC052424.1 | - | 4 | 2296 | 6835200 | ||
BC054843.1 | - | 4 | 2272 |