Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pole4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025882.3 | NP_080158.1 | 2, 3 | 325 | ||
GenBank mRNA | AK010135.1 | - | 2, 3 | 320 | ||
AK010300.1 | - | 2, 3 | 326 | |||
AK011595.1 | - | 2, 3 | 337 | |||
AK145365.1 | - | 2, 3 | 331 | |||
AK160582.1 | - | 2, 3 | 337 | |||
AK167631.1 | - | 2, 3 | 315 | |||
BC023189.1 | - | 2, 3 | 303 | 4952949 |